Product Details

SNP ID
rs3212994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113122816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGCGTGGACTTTGCTCCAGCAGC[C/T]TGTCCCAGTGAGGACAGGGACACAG
Phenotype
MIM: 613872 MIM: 613878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F10 PubMed Links

Gene Details

Gene
F10
Gene Name
coagulation factor X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000504.3 18 UTR 5 NP_000495.1
NM_001312674.1 18 UTR 5 NP_001299603.1
NM_001312675.1 18 UTR 5 NP_001299604.1
Gene
F10-AS1
Gene Name
F10 antisense RNA 1
There are no transcripts associated with this gene.

Gene
F7
Gene Name
coagulation factor VII
There are no transcripts associated with this gene.

View Full Product Details