Product Details

SNP ID
rs4958303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:152392112 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGAGCCATTCTACCTCTCTAAT[A/T]TCATATGAGAAGGCATACATTATGG
Phenotype
MIM: 605108
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NMUR2 PubMed Links

Gene Details

Gene
NMUR2
Gene Name
neuromedin U receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020167.4 1493 UTR 3 NP_064552.3
XM_011537670.2 1493 Intron XP_011535972.1

View Full Product Details