Product Details

SNP ID
hCV32407220
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.22:42126667 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CCGGGGCTGTCCAGTGGGCAC[AGTGGGCAC/-]CGAGAAGCTGAAGTGCTGCAG
Phenotype
MIM: 124030
Polymorphism
AGTGGGCAC/-, Insertion/deletion
Allele Nomenclature
CYP2D6*18,g.4125_4133dupGTGCCCACT
Literature Links
CYP2D6 PubMed Links
Additional Information
The CYP2D6 gene exhibits copy number variation. Individuals may carry deletion alleles or extra copies of CYP2D6. CYP2D6 SNP genotyping assays run on samples lacking CYP2D6 genes will not amplify, homozygous samples having 1 or more gene copies typically cluster together, and heterozygous samples with more than 2 copies may run between the 2 copy heterozygous and homozygous genotype clusters. In addition, some CYP2D6 alleles contain CYP2D7 pseudogene sequences. For accurate CYP2D6 genotype analysis, copy number analysis must be done. For more information, refer to the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 2 Copy Number Variation section.

Gene Details

Gene
CYP2D6
Gene Name
cytochrome P450 family 2 subfamily D member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000106.5 1338 Amino Acid InDel TCG,TCGTGCCCACTG S467SCPL NP_000097.3
NM_001025161.2 1338 Amino Acid InDel TCG,TCGTGCCCACTG S416SCPL NP_001020332.2
Gene
LOC102723722
Gene Name
uncharacterized LOC102723722
There are no transcripts associated with this gene.

Gene
NDUFA6-AS1
Gene Name
NDUFA6 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details