Product Details

SNP ID
rs16942050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112422309 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTACTGATATCTCTCAGCATCA[C/T]AGGTAGGAAAACTAAGGCATTCAGC
Phenotype
MIM: 176876 MIM: 603703
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN11 PubMed Links

Gene Details

Gene
PTPN11
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719526.2 Intron XP_006719589.1
XM_011538613.2 Intron XP_011536915.1
XM_017019722.1 Intron XP_016875211.1
Gene
RPL6
Gene Name
ribosomal protein L6
There are no transcripts associated with this gene.

View Full Product Details