Product Details

SNP ID
rs17578859
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136984718 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGGCTCTCGCTCCTCCAGCCATG[A/G]CCCTGAGTGACATCCGAGTGGCCTT
Phenotype
MIM: 616315 MIM: 606533 MIM: 176803
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
There are no transcripts associated with this gene.

Gene
LCNL1
Gene Name
lipocalin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207510.3 796 Missense Mutation ACC,GCC T68A NP_997393.3
Gene
PTGDS
Gene Name
prostaglandin D2 synthase
There are no transcripts associated with this gene.

View Full Product Details