Product Details
- SNP ID
-
rs17832339
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:90014262 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCACAGTACCATGGATCAACTCTT[G/T]ACTCACAAATTGAATGAAGGGATGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3174
PubMed Links
Gene Details
- Gene
- MIR3174
- Gene Name
- microRNA 3174
There are no transcripts associated with this gene.
- Gene
- ZNF710
- Gene Name
- zinc finger protein 710
View Full Product Details