Product Details
- SNP ID
-
rs16855035
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108958975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATACCATCTCTGACTTAATTTTCC[C/G]AAGTCTTTTCATTTTTTTCTAAAGG
- Phenotype
-
MIM: 605128
MIM: 603205
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GUCA1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2197737] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GUCA1C
- Gene Name
- guanylate cyclase activator 1C
There are no transcripts associated with this gene.
- Gene
- MORC1
- Gene Name
- MORC family CW-type zinc finger 1
View Full Product Details