Product Details

SNP ID
rs16838982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127920229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCACCAAATGCTCATACTTACTT[C/T]GTTGAAATCGATCATCAGTATTTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD12 PubMed Links

Gene Details

Gene
KBTBD12
Gene Name
kelch repeat and BTB domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207335.2 Intron NP_997218.2
XM_005247167.1 Intron XP_005247224.1
XM_005247168.4 Intron XP_005247225.1
XM_011512490.2 Intron XP_011510792.1
XM_011512491.2 Intron XP_011510793.1
XM_011512492.2 Intron XP_011510794.1

View Full Product Details