Product Details

SNP ID
rs16876100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73552472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATAAAGTGAATGACTTAATACA[C/T]GAGTGAAGATGACTAGGGTAGAATA
Phenotype
MIM: 605787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 1784 UTR 3 NP_075526.1
XM_005248560.2 1784 UTR 3 XP_005248617.1
XM_017009678.1 1784 UTR 3 XP_016865167.1

View Full Product Details