Product Details

SNP ID
rs17022564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99322568 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCGGAAGAGACTTACAAGAATC[A/G]GAAGTGCACACATGGTGCCATATTT
Phenotype
MIM: 612564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNDC9 PubMed Links

Gene Details

Gene
TXNDC9
Gene Name
thioredoxin domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005783.3 Intron NP_005774.2
XM_017003147.1 Intron XP_016858636.1

View Full Product Details