Product Details

SNP ID
rs34236746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:43699073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGATGGACTTTGAGATCTACTTG[C/T]AAGTACACAGGCTGAAGTTGAATAT
Phenotype
MIM: 604780 MIM: 613726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD5 PubMed Links

Gene Details

Gene
ABHD5
Gene Name
abhydrolase domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016006.4 Intron NP_057090.2
XM_011533779.2 Intron XP_011532081.1
XM_011533780.1 Intron XP_011532082.1
Gene
ANO10
Gene Name
anoctamin 10
There are no transcripts associated with this gene.

View Full Product Details