Product Details

SNP ID
rs17562811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35972858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGCGGGTAGGTGTTCCTTTGCT[C/T]CTCACAAGTCTCTTTCTCTTTTACA
Phenotype
MIM: 610347
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C18orf21 PubMed Links

Gene Details

Gene
C18orf21
Gene Name
chromosome 18 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201474.1 234 Intron NP_001188403.1
NM_001201475.1 234 UTR 5 NP_001188404.1
NM_001201476.1 234 Intron NP_001188405.1
NM_031446.4 234 Intron NP_113634.3
XM_005258364.4 234 Intron XP_005258421.1
Gene
RPRD1A
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
There are no transcripts associated with this gene.

View Full Product Details