Product Details

SNP ID
rs17182621
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73854013 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGGGATGAATTTAACAATCTAC[A/C]CTTCTTTCAGATCTTGGAACTTGAT
Phenotype
MIM: 608642
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PTGR2 PubMed Links
Additional Information
For this assay, SNP(s) [rs79188481] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PTGR2
Gene Name
prostaglandin reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146154.1 Intron NP_001139626.1
NM_001146155.1 Intron NP_001139627.1
NM_152444.2 Intron NP_689657.1

View Full Product Details