Product Details

SNP ID
rs8025811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:62164746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGACAGCGCTGCCTGCGAGCGC[A/G]GGTCCCAGTCCGTGCGGCCGGCGTC
Phenotype
MIM: 610344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD4B PubMed Links

Gene Details

Gene
C2CD4B
Gene Name
C2 calcium dependent domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007595.2 709 Missense Mutation CCG,CTG P80L NP_001007596.2
XM_017022170.1 709 Missense Mutation CCG,CTG P80L XP_016877659.1

View Full Product Details