Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001241.3 | Intron | NP_001232.1 | |||
NM_001320748.1 | Intron | NP_001307677.1 | |||
NM_001320749.1 | Intron | NP_001307678.1 | |||
NM_058241.2 | Intron | NP_490595.1 | |||
XM_017005227.1 | Intron | XP_016860716.1 | |||
XM_017005228.1 | Intron | XP_016860717.1 | |||
XM_017005229.1 | Intron | XP_016860718.1 | |||
XM_017005230.1 | Intron | XP_016860719.1 | |||
XM_017005231.1 | Intron | XP_016860720.1 | |||
XM_017005232.1 | Intron | XP_016860721.1 | |||
XM_017005233.1 | Intron | XP_016860722.1 |