Product Details
- SNP ID
-
rs16877194
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:6314312 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCAGGAGCTCCTCTGTGCTCAAT[C/T]CTGAGATCATGAGAGCCAGGATGGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ33360
PubMed Links
Gene Details
- Gene
- FLJ33360
- Gene Name
- FLJ33360 protein
There are no transcripts associated with this gene.
View Full Product Details