Product Details

SNP ID
rs16845326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70657937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCGTGATGTCCCATGAAAAATTAG[A/T]TGATTGTCAACGAGTTCAGGTGATT
Phenotype
MIM: 147790
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JCHAIN PubMed Links

Gene Details

Gene
JCHAIN
Gene Name
joining chain of multimeric IgA and IgM
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144646.3 Intron NP_653247.1
XM_006714211.2 Intron XP_006714274.1
XM_006714212.2 Intron XP_006714275.1
XM_011531925.1 Intron XP_011530227.1
XM_011531926.1 Intron XP_011530228.1

View Full Product Details