Product Details

SNP ID
rs16845387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70688761 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGTCCCACGCTCACCGACGAAAA[C/T]GGAGATGATTTAGGATTGCCACCCT
Phenotype
MIM: 611614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UTP3 PubMed Links

Gene Details

Gene
UTP3
Gene Name
UTP3, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020368.2 283 Silent Mutation AAC,AAT N28N NP_065101.1

View Full Product Details