Product Details
- SNP ID
-
rs17695522
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:27287222 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAATGATATTTAGGTGATAAGCAA[C/T]GACAAGCTTCCATGTCTATATTCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EQTN
PubMed Links
Gene Details
- Gene
- EQTN
- Gene Name
- equatorin
There are no transcripts associated with this gene.
- Gene
- LINC00032
- Gene Name
- long intergenic non-protein coding RNA 32
There are no transcripts associated with this gene.
View Full Product Details