Product Details

SNP ID
rs17169429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135936789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTTTACTAGATCGGGCCTGTGC[A/G]TCTTCTGTATGTAGGAGGTGGAATG
Phenotype
MIM: 609087
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL21 PubMed Links

Gene Details

Gene
FBXL21
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012159.4 677 Silent Mutation GCA,GCG A65A NP_036291.2

View Full Product Details