Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256182.1 | 8871 | Intron | NP_001243111.1 | ||
NM_001256183.1 | 8871 | Intron | NP_001243112.1 | ||
NM_013275.5 | 8871 | Intron | NP_037407.4 | ||
XM_006721181.1 | 8871 | Intron | XP_006721244.1 | ||
XM_006721184.2 | 8871 | Intron | XP_006721247.1 | ||
XM_011523051.2 | 8871 | Intron | XP_011521353.1 | ||
XM_011523053.2 | 8871 | Intron | XP_011521355.1 | ||
XM_011523054.2 | 8871 | Intron | XP_011521356.1 | ||
XM_011523055.2 | 8871 | Intron | XP_011521357.1 | ||
XM_011523056.2 | 8871 | Intron | XP_011521358.1 | ||
XM_011523057.2 | 8871 | UTR 3 | XP_011521359.1 | ||
XM_017023182.1 | 8871 | Intron | XP_016878671.1 | ||
XM_017023183.1 | 8871 | Intron | XP_016878672.1 | ||
XM_017023184.1 | 8871 | Intron | XP_016878673.1 | ||
XM_017023185.1 | 8871 | Intron | XP_016878674.1 | ||
XM_017023186.1 | 8871 | Intron | XP_016878675.1 | ||
XM_017023187.1 | 8871 | Intron | XP_016878676.1 |