Product Details

SNP ID
rs17851290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119932116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAGAAGAACCTACTTGTTCGAT[A/C]CTTCCAGGGATTTTCCAGAAACTCT
Phenotype
MIM: 300766
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NKAP PubMed Links

Gene Details

Gene
NKAP
Gene Name
NFKB activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024528.3 1005 Missense Mutation GAT,TAT D280Y NP_078804.2
XM_017029842.1 1005 Missense Mutation GAT,TAT D181Y XP_016885331.1

View Full Product Details