Product Details

SNP ID
rs16943885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74447429 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCCAGTCACCACAGAGAAGATT[C/T]AACAAGTAAAAACCTGCAAACTCTT
Phenotype
MIM: 600753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLG1 PubMed Links

Gene Details

Gene
GLG1
Gene Name
golgi glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145666.1 8256 UTR 3 NP_001139138.1
NM_001145667.1 8256 UTR 3 NP_001139139.1
NM_012201.5 8256 UTR 3 NP_036333.2

View Full Product Details