Product Details

SNP ID
rs16972190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19558206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCAGAGGAGAAATGAGTTTCTGT[C/T]ATTAAATTTACATTTATCAAGTCCT
Phenotype
MIM: 609544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf62 PubMed Links

Gene Details

Gene
C16orf62
Gene Name
chromosome 16 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300743.1 Intron NP_001287672.1
NM_020314.5 Intron NP_064710.4
XM_005255435.3 Intron XP_005255492.1
XM_006721065.3 Intron XP_006721128.1
XM_011545896.2 Intron XP_011544198.1
XM_011545897.2 Intron XP_011544199.1
Gene
CCP110
Gene Name
centriolar coiled-coil protein 110
There are no transcripts associated with this gene.

View Full Product Details