Product Details

SNP ID
rs35904671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24009481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCAATGATCATTTTAGAGGGAGC[A/G]TAACAACCGCCTGACTGTCGCCTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD2B PubMed Links

Gene Details

Gene
MFSD2B
Gene Name
major facilitator superfamily domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080473.1 460 Intron NP_001073942.1
XM_005264301.4 460 UTR 5 XP_005264358.1
XM_017004063.1 460 UTR 5 XP_016859552.1
XM_017004064.1 460 UTR 5 XP_016859553.1
XM_017004065.1 460 UTR 5 XP_016859554.1
XM_017004066.1 460 UTR 5 XP_016859555.1
XM_017004067.1 460 UTR 5 XP_016859556.1
XM_017004068.1 460 UTR 5 XP_016859557.1
XM_017004069.1 460 UTR 5 XP_016859558.1
XM_017004070.1 460 UTR 5 XP_016859559.1
Gene
UBXN2A
Gene Name
UBX domain protein 2A
There are no transcripts associated with this gene.

View Full Product Details