Product Details

SNP ID
rs17749735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53157749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAAGGGAGGGAGCTCAAAAGA[C/T]ATAAAGAAGTCTATGTTTTTTTTGT
Phenotype
MIM: 616569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSAD PubMed Links

Gene Details

Gene
CSAD
Gene Name
cysteine sulfinic acid decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244705.1 2129 UTR 3 NP_001231634.1
NM_001244706.1 2129 Intron NP_001231635.1
NM_015989.4 2129 UTR 3 NP_057073.4
XM_011538442.2 2129 UTR 3 XP_011536744.2
XM_011538446.2 2129 UTR 3 XP_011536748.1
XM_011538449.1 2129 UTR 3 XP_011536751.1
XM_011538451.1 2129 UTR 3 XP_011536753.1
XM_017019416.1 2129 Intron XP_016874905.1
XM_017019417.1 2129 UTR 3 XP_016874906.1
XM_017019418.1 2129 UTR 3 XP_016874907.1
XM_017019419.1 2129 UTR 3 XP_016874908.1

View Full Product Details