Product Details

SNP ID
rs17539401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15217954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGATGAGCCTTACTTATTTGCA[A/G]TGTAAAATCTGTATGCTGAGTGGCT
Phenotype
MIM: 606400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN7 PubMed Links

Gene Details

Gene
CAPN7
Gene Name
calpain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014296.2 Intron NP_055111.1
XM_011533555.2 Intron XP_011531857.1
XM_011533556.2 Intron XP_011531858.1
XM_011533557.2 Intron XP_011531859.1
XM_017006043.1 Intron XP_016861532.1
XM_017006044.1 Intron XP_016861533.1
XM_017006045.1 Intron XP_016861534.1

View Full Product Details