Product Details

SNP ID
rs17030552
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33388966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTCAAATCAGGACACCTGGGTA[C/T]ATGGACAAAGATATCCTACAATTAA
Phenotype
MIM: 605652 MIM: 609784
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FBXL2 PubMed Links

Gene Details

Gene
FBXL2
Gene Name
F-box and leucine rich repeat protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171713.1 3518 Intron NP_001165184.1
NM_012157.3 3518 Intron NP_036289.3
XM_005265014.2 3518 Intron XP_005265071.1
XM_005265015.3 3518 Intron XP_005265072.1
XM_005265016.2 3518 Intron XP_005265073.1
XM_005265017.3 3518 Intron XP_005265074.1
XM_011533559.2 3518 Intron XP_011531861.1
XM_017006092.1 3518 Intron XP_016861581.1
XM_017006093.1 3518 Intron XP_016861582.1
XM_017006094.1 3518 Intron XP_016861583.1
Gene
UBP1
Gene Name
upstream binding protein 1 (LBP-1a)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128160.1 3518 UTR 3 NP_001121632.1
NM_001128161.1 3518 UTR 3 NP_001121633.1
NM_014517.4 3518 UTR 3 NP_055332.3

View Full Product Details