Product Details

SNP ID
rs17544587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169645891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGACTTTTGAAAATTATATTTTG[A/G]TCCCTGAGAGTTATAAAAAGGGAGC
Phenotype
MIM: 606093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC173 PubMed Links

Gene Details

Gene
CCDC173
Gene Name
coiled-coil domain containing 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085447.1 1624 Missense Mutation CCA,TCA P537S NP_001078916.1
XM_011510589.1 1624 Missense Mutation CCA,TCA P513S XP_011508891.1
XM_011510590.1 1624 Missense Mutation CCA,TCA P456S XP_011508892.1
Gene
PPIG
Gene Name
peptidylprolyl isomerase G
There are no transcripts associated with this gene.

View Full Product Details