Product Details
- SNP ID
-
rs778369
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:232879421 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGCTGGCCTGTGCTTCCCAGAG[C/G]CCCCCCCCCCCCACCTTCTGTCGGG
- Phenotype
-
MIM: 605991
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf82
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs570180043] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf82
- Gene Name
- chromosome 2 open reading frame 82
- Gene
- NGEF
- Gene Name
- neuronal guanine nucleotide exchange factor
View Full Product Details