Product Details

SNP ID
rs16823097
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:184395379 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAATATAATGAGGAAGTATAGT[C/T]TGGAATGAGAAAAGATAATATAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf21 PubMed Links

Gene Details

Gene
C1orf21
Gene Name
chromosome 1 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030806.3 Intron NP_110433.1

View Full Product Details