Product Details

SNP ID
rs17167929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:14148044 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATAAACTGATACAATCATTTAC[C/T]TCCTTCAAGTGTACTTAACTGATCC
Phenotype
MIM: 604070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKB PubMed Links

Gene Details

Gene
DGKB
Gene Name
diacylglycerol kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004080.2 4214 UTR 3 NP_004071.1
NM_145695.2 4214 Intron NP_663733.1
XM_005249628.2 4214 UTR 3 XP_005249685.1
XM_005249629.2 4214 UTR 3 XP_005249686.1
XM_005249630.2 4214 UTR 3 XP_005249687.1
XM_005249631.3 4214 UTR 3 XP_005249688.1
XM_011515153.2 4214 UTR 3 XP_011513455.1
XM_011515154.2 4214 UTR 3 XP_011513456.1
XM_011515156.2 4214 UTR 3 XP_011513458.1
XM_011515157.2 4214 UTR 3 XP_011513459.1
XM_011515158.2 4214 UTR 3 XP_011513460.1
XM_017011783.1 4214 UTR 3 XP_016867272.1
XM_017011784.1 4214 UTR 3 XP_016867273.1
XM_017011785.1 4214 UTR 3 XP_016867274.1
XM_017011786.1 4214 UTR 3 XP_016867275.1
XM_017011787.1 4214 UTR 3 XP_016867276.1
XM_017011788.1 4214 UTR 3 XP_016867277.1
XM_017011789.1 4214 UTR 3 XP_016867278.1
XM_017011790.1 4214 Intron XP_016867279.1
XM_017011791.1 4214 Intron XP_016867280.1
XM_017011792.1 4214 Intron XP_016867281.1

View Full Product Details