Product Details

SNP ID
rs16878759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16463459 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTTAAATATTGTGTATAATGCA[C/T]ATGCTCTCGGTAATATTTTATTTCA
Phenotype
MIM: 609675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOSTDC1 PubMed Links

Gene Details

Gene
SOSTDC1
Gene Name
sclerostin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015464.2 Intron NP_056279.1
XM_017011943.1 Intron XP_016867432.1

View Full Product Details