Product Details

SNP ID
rs17066609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45333787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTTCCCACTAGTGAAATCCAAC[A/G]GTAGTCAAGTTTACCACTATCAGGC
Phenotype
MIM: 600763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORA31 PubMed Links

Gene Details

Gene
SNORA31
Gene Name
small nucleolar RNA, H/ACA box 31
There are no transcripts associated with this gene.

Gene
TPT1
Gene Name
tumor protein, translationally-controlled 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286272.1 4498 UTR 3 NP_001273201.1
NM_001286273.1 4498 UTR 3 NP_001273202.1
NM_003295.3 4498 UTR 3 NP_003286.1
Gene
TPT1-AS1
Gene Name
TPT1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details