Product Details

SNP ID
rs17004600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43670062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGCCAGTCCCCCGATACACTGA[C/T]GCACACTGACAGTCATGCTTCAAGG
Phenotype
MIM: 610654
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP1B PubMed Links

Gene Details

Gene
RRP1B
Gene Name
ribosomal RNA processing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015056.2 Intron NP_055871.1

View Full Product Details