Product Details

SNP ID
hCV33410087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:36787404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAGAAATGTTGCTTTCAGCCCA[A/G]ACCTTTGTGGGGCAAGTGTTGGTAA
Phenotype
MIM: 615215
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNU1 PubMed Links

Gene Details

Gene
KCNU1
Gene Name
potassium calcium-activated channel subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031836.2 381 Silent Mutation CAA,CAG Q98Q NP_001027006.2

View Full Product Details