Product Details

SNP ID
rs16890177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37823325 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAAAATTTTATGCAGCTTTGCA[C/G]TCCTTGGAAGCTCTAGGTCTAAAGT
Phenotype
MIM: 607455
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZFAND3 PubMed Links

Gene Details

Gene
ZFAND3
Gene Name
zinc finger AN1-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021943.2 Intron NP_068762.1
XM_005249269.1 Intron XP_005249326.1
XM_011514790.1 Intron XP_011513092.1
XM_017011171.1 Intron XP_016866660.1

View Full Product Details