Product Details
- SNP ID
-
rs16890915
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:38677131 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGCTTGAAAACCAGAATGACTGA[A/G]CAGTTAGGTGAAAAGGAACAGCTGA
- Phenotype
-
MIM: 138750
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLO1
PubMed Links
Gene Details
- Gene
- GLO1
- Gene Name
- glyoxalase I
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006708.2 |
841 |
UTR 3 |
|
|
NP_006699.2 |
View Full Product Details