Product Details
- SNP ID
-
rs17131140
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:90713638 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGGGCATTTCCAACCGCACTTATG[A/G]ACGATTAGAACACAAACGGGAGAAA
- Phenotype
-
MIM: 605212
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BARHL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111253587] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BARHL2
- Gene Name
- BarH like homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020063.1 |
|
Intron |
|
|
NP_064447.1 |
View Full Product Details