Product Details
- SNP ID
-
rs17003846
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101080877 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCAGACCCTAGTCAGCTGCTTCA[A/G]AGGCCCCCCCAAGCTGCACCCTAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM35A
PubMed Links
Gene Details
- Gene
- TMEM35A
- Gene Name
- transmembrane protein 35A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021637.2 |
|
Intron |
|
|
NP_067650.1 |
View Full Product Details