Product Details
- SNP ID
-
rs16839255
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
33
- Location
-
Chr.1:157832886 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTCTCACTTTATGTTTACTATGTT[C/T]AATATAAAAATGTTGGTGAAATAAT
- Phenotype
-
MIM: 602592
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD5L
PubMed Links
Gene Details
- Gene
- CD5L
- Gene Name
- CD5 molecule like
View Full Product Details