Product Details

SNP ID
rs17002590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54749729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCCTGACTCTATGTCTCTGCATG[C/T]CTATGTGTTTGTATACCCTTGTATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITIH6 PubMed Links

Gene Details

Gene
ITIH6
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198510.2 4138 UTR 3 NP_940912.1

View Full Product Details