Product Details

SNP ID
rs16984267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52969304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACTTCATTATGCATTCTCCAAT[A/G]ATTTGCAATGGTTGTAGCATTACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF702P PubMed Links

Gene Details

Gene
ZNF702P
Gene Name
zinc finger protein 702, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF816
Gene Name
zinc finger protein 816
There are no transcripts associated with this gene.

Gene
ZNF816-ZNF321P
Gene Name
ZNF816-ZNF321P readthrough
There are no transcripts associated with this gene.

View Full Product Details