Product Details

SNP ID
rs17128917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93182605 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCCCTCAGAGCACTGCACACAG[A/G]TAGTGAAAGAACTTGTGTACAATAA
Phenotype
MIM: 609485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOAP1 PubMed Links

Gene Details

Gene
MOAP1
Gene Name
modulator of apoptosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022151.4 2008 UTR 3 NP_071434.2
Gene
TMEM251
Gene Name
transmembrane protein 251
There are no transcripts associated with this gene.

View Full Product Details