Product Details

SNP ID
rs34870356
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:180200025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGGGGACCCAGCCAGCCTTGA[A/C]CCTGGAGGAAAGTCAGGTGGGCTCA
Phenotype
MIM: 603120
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLJ23867 PubMed Links

Gene Details

Gene
FLJ23867
Gene Name
uncharacterized protein FLJ23867
There are no transcripts associated with this gene.

Gene
QSOX1
Gene Name
quiescin sulfhydryl oxidase 1
There are no transcripts associated with this gene.

View Full Product Details