Product Details

SNP ID
rs16974177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89789353 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTGTCAAGCTTCTAATTCAAGC[A/G]TATATATCAATTGTAAAAAGACCTT
Phenotype
MIM: 151530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANPEP PubMed Links

Gene Details

Gene
ANPEP
Gene Name
alanyl aminopeptidase, membrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001150.2 Intron NP_001141.2
XM_005254892.4 Intron XP_005254949.1
XM_011521473.1 Intron XP_011519775.1

View Full Product Details