Product Details

SNP ID
rs17101052
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:77905306 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCATGTTGTATTTATACAATATCC[C/T]TGCATAATGATAATCATAATGATAG
Phenotype
MIM: 613121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEXN PubMed Links

Gene Details

Gene
NEXN
Gene Name
nexilin F-actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172309.1 Intron NP_001165780.1
NM_144573.3 Intron NP_653174.3
XM_005271322.3 Intron XP_005271379.1
XM_005271323.3 Intron XP_005271380.1
XM_005271324.4 Intron XP_005271381.1
XM_005271325.3 Intron XP_005271382.1
XM_005271326.3 Intron XP_005271383.1
XM_005271327.3 Intron XP_005271384.1

View Full Product Details