Product Details

SNP ID
rs17102355
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:46638198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCACACTACAACACTGCTGTTCAA[A/T]CTATTCTGTAGAGTCCTAGGTGCTC
Phenotype
MIM: 608917
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042546.2 Intron NP_001036011.2
NM_001243728.1 Intron NP_001230657.1
NM_001256418.1 Intron NP_001243347.1
NM_022745.4 Intron NP_073582.3

View Full Product Details