Product Details

SNP ID
rs10907134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:123865197 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGACATTGTGTGAGTAGATATGA[A/G]GAGACAAACAGTAGTGATAAGATAA
Phenotype
MIM: 300079
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XIAP PubMed Links

Gene Details

Gene
XIAP
Gene Name
X-linked inhibitor of apoptosis
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167.3 Intron NP_001158.2
NM_001204401.1 Intron NP_001191330.1
XM_006724754.2 Intron XP_006724817.1
XM_011531329.2 Intron XP_011529631.1

View Full Product Details