Product Details
- SNP ID
-
rs17842564
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:121539177 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTGGGGGAGCATTTTTGTGACTT[A/T]GTGCAGCTTTTGTTTGTTCCCAAAT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EMBP1
PubMed Links
Gene Details
- Gene
- EMBP1
- Gene Name
- embigin pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details